Merck
  • Interaction of metabolites with R. rhodochrous during the biodegradation of di-ester plasticizers.
Interaction of metabolites with R. rhodochrous during the biodegradation of di-ester plasticizers.
Chemosphere (2006-06-17)
Sandro Nalli, David G Cooper, Jim A Nicell
ABSTRACT
The commonly used plasticizers di-ethylhexyl phthalate (DEHP) and di-ethylhexyl adipate (DEHA) are known to partially degrade in the presence of soil microorganisms, such as Rhodococcus rhodochrous, releasing persistent and toxic metabolites. The metabolites adipic acid and 2-ethylhexanol were both shown to inhibit growth of the degrading microbe. 2-Ethylhexanol enhanced the activity of ethanol dehydrogenase - an enzyme involved in its metabolism - but the activity of this enzyme was inhibited by adipic acid. The metabolite usually seen in the highest concentrations - 2-ethylhexanoic acid - did not exhibit any evidence of inhibition. It was shown that the high concentration of this metabolite was due to the inability of R. rhodochrous to degrade it. Comparisons with other small carboxylic acids supported the argument that the ethyl branch was the reason for the resistance of 2-ethylhexanoic acid to degradation. The hydrophobicity of the cell surface was shown to be a factor in plasticizer degradation. The primary carbon source could be either water-soluble or hydrophobic and a hydrophobic substrate led to a cell surface that attracted the plasticizer and facilitated degradation. The most hydrophobic of the plasticizers, DEHP, was particularly sensitive to this effect.
Sigma-Aldrich
Titanium(IV) 2-ethylhexyloxide, 95%
Sigma-Aldrich
2-Ethyl-1-hexanol, puriss., ≥99.0% (GC)
Sigma-Aldrich
2-Ethyl-1-hexanol, ≥99.6%
Supelco
2-Ethyl-1-hexanol, analytical standard
Sigma-Aldrich
2-Ethyl-1-hexanol, ≥99%, FG