Merck
  • Effects of prenatal exposure to the environmental pollutant 2-bromopropane on embryo-fetal development in rats.
Effects of prenatal exposure to the environmental pollutant 2-bromopropane on embryo-fetal development in rats.
Toxicology (2004-03-24)
Jong-Choon Kim, Sung-Ho Kim, Dong-Ho Shin, Tae-Ho Ahn, Hyoung-Chin Kim, Yun-Bae Kim, Cheng-Zhe Jiang, Junghee Han, Moon-Koo Chung
ABSTRACT
2-Bromopropane (2-BP), a halogenated propane analogue, is a substitute for chlorofluorocarbones. The present study was carried out to investigate the potential adverse effects of 2-BP on pregnant dams and embryo-fetal development after maternal exposure on gestational days (GD) 6 through 19 in Sprague-Dawley rats. The test chemical was administered subcutaneously to pregnant rats at dose levels of 0, 250, 500, and 1000 mg/kg per day. All dams were subjected to caesarean section on GD 20 and their fetuses were examined for external, visceral and skeletal abnormalities. In the 1000 mg/kg group, maternal toxicity included an increase in the incidence of abnormal clinical signs, a suppression in the body weight and body weight gain, and a decrease in the food intake. Developmental toxicity included an increase in the fetal deaths, a decrease in the litter size, and a reduction in the fetal body weight. In addition, an increase in the incidence of fetal external, visceral, and skeletal abnormalities was seen. In the 500 mg/kg group, minimal developmental toxicity including decreased fetal body weight and increased fetal ossification delay was observed. There were no adverse effects on either pregnant dams or embryo-fetal development in the 250 mg/kg group. These findings suggest that a 14-day subcutaneous dose of 2-BP is embryotoxic and teratogenic at a maternally toxic dose (i.e., 1000 mg/kg per day) and is minimally embryotoxic at a nonmaternally toxic dose (i.e., 500 mg/kg per day) in Sprague-Dawley rats. In the present experimental conditions, the no-observed-adverse-effect level (NOAEL) of 2-BP is considered to be 500 mg/kg per day for dams and 250 mg/kg per day for embryo-fetal development.
Sigma-Aldrich
2-Bromopropane, 99%