Merck
  • An anti-PD-1-GITR-L bispecific agonist induces GITR clustering-mediated T cell activation for cancer immunotherapy.
An anti-PD-1-GITR-L bispecific agonist induces GITR clustering-mediated T cell activation for cancer immunotherapy.
Nature cancer (2022-03-09)
Sarah Chan, Nicole Belmar, Sun Ho, Bryan Rogers, Marcia Stickler, Michelle Graham, Eileen Lee, Ninian Tran, Dong Zhang, Priyanka Gupta, Mien Sho, Tracy MacDonough, Andrew Woolley, Han Kim, Hong Zhang, Wei Liu, Pingping Zheng, Zoltan Dezso, Kyle Halliwill, Michele Ceccarelli, Susan Rhodes, Archana Thakur, Charles M Forsyth, Mengli Xiong, Siu Sze Tan, Ramesh Iyer, Marc Lake, Enrico Digiammarino, Li Zhou, Lance Bigelow, Kenton Longenecker, Russell A Judge, Cassie Liu, Max Trumble, Jonathan P Remis, Melvin Fox, Belinda Cairns, Yoshiko Akamatsu, Diane Hollenbaugh, Fiona Harding, Hamsell M Alvarez
ABSTRACT
Costimulatory receptors such as glucocorticoid-induced tumor necrosis factor receptor-related protein (GITR) play key roles in regulating the effector functions of T cells. In human clinical trials, however, GITR agonist antibodies have shown limited therapeutic effect, which may be due to suboptimal receptor clustering-mediated signaling. To overcome this potential limitation, a rational protein engineering approach is needed to optimize GITR agonist-based immunotherapies. Here we show a bispecific molecule consisting of an anti-PD-1 antibody fused with a multimeric GITR ligand (GITR-L) that induces PD-1-dependent and FcγR-independent GITR clustering, resulting in enhanced activation, proliferation and memory differentiation of primed antigen-specific GITR+PD-1+ T cells. The anti-PD-1-GITR-L bispecific is a PD-1-directed GITR-L construct that demonstrated dose-dependent, immunologically driven tumor growth inhibition in syngeneic, genetically engineered and xenograft humanized mouse tumor models, with a dose-dependent correlation between target saturation and Ki67 and TIGIT upregulation on memory T cells. Anti-PD-1-GITR-L thus represents a bispecific approach to directing GITR agonism for cancer immunotherapy.
Sigma-Aldrich
Lectin from Phaseolus vulgaris (red kidney bean), Phytohemagglutinin PHA-M, lyophilized powder, BioReagent, suitable for cell culture
Pricing and availability is not currently available.