Merck
  • Male-derived copulatory plugs enhance implantation success in female Mus musculus.
Male-derived copulatory plugs enhance implantation success in female Mus musculus.
Biology of reproduction (2020-12-24)
Michael Lough-Stevens, Caleb R Ghione, Matthew Urness, Adelaide Hobbs, Colleen M Sweeney, Matthew D Dean
ABSTRACT
Among a wide diversity of sexually reproducing species, male ejaculates coagulate to form what has been termed a copulatory plug. A number of functions have been attributed to copulatory plugs, including the inhibition of female remating and the promotion of ejaculate movement. Here we demonstrate that copulatory plugs also influence the likelihood of implantation, which occurs roughly 4 days after copulation in mice. Using a bead transfer method to control for differences in ejaculate retention and fertilization rates, we show that implantation rates significantly drop among females mated to genetically engineered males incapable of forming plugs (because they lack functional transglutaminase 4, the main enzyme responsible for its formation). Surprisingly, this result does not correlate with differences in circulating progesterone levels among females, an important hormone involved in implantation. We discuss three models that connect male-derived copulatory plugs to implantation success, including the hypothesis that plugs contribute to a threshold amount of stimulation required for females to become receptive to implantation.
Sigma-Aldrich
Hyaluronidase from bovine testes, Type IV-S, powder, suitable for mouse embryo cell culture, 750-3000 units/mg solid
Sigma-Aldrich
Concanavalin A from Canavalia ensiformis (Jack bean), Sepharose conjugate, buffered aqueous suspension