- 1,3,5-Triazine Nitrogen Mustards with Different Peptide Group as Innovative Candidates for AChE and BACE1 Inhibitors.
1,3,5-Triazine Nitrogen Mustards with Different Peptide Group as Innovative Candidates for AChE and BACE1 Inhibitors.
Molecules (Basel, Switzerland) (2021-07-03)
Dawid Maliszewski, Agnieszka Wróbel, Beata Kolesińska, Justyna Frączyk, Danuta Drozdowska
PMID34203347
ABSTRACT
A series of new analogs of nitrogen mustards (4a-4h) containing the 1,3,5-triazine ring substituted with dipeptide residue were synthesized and evaluated for the inhibition of both acetylcholinesterase (AChE) and β-secretase (BACE1) enzymes. The AChE inhibitory activity studies were carried out using Ellman's colorimetric method, and the BACE1 inhibitory activity studies were carried out using fluorescence resonance energy transfer (FRET). All compounds displayed considerable AChE and BACE1 inhibition. The most active against both AChE and BACE1 enzymes were compounds A and 4a, with an inhibitory concentration of AChE IC50 = 0.051 µM; 0.055 µM and BACE1 IC50 = 9.00 µM; 11.09 µM, respectively.
Sigma-Aldrich
Acetylcholinesterase from Electrophorus electricus (electric eel), Type VI-S, lyophilized powder, 200-1,000 units/mg protein
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
β-Secretase (BACE1) Activity Detection Kit (Fluorescent), 1 kit sufficient for 250 reactions
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acetylcholinesterase Inhibitor Screening Kit, sufficient for 100 colorimetric tests