
Torsemide Related Compound E

Synonym(s):
4-(3-Methylphenyl)-2H-pyrido[4,3-e]-1,2,4-thiadiazin-3(4H)-one 1,1-dioxide, 4-m-Tolyl-2H-pyrido[4,3-e][1,2,4]thiadiazin-3(4H)-one 1,1-dioxide
Empirical Formula (Hill Notation):
C13H11N3O3S
CAS Number:
Molecular Weight:
289.31
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2