
Procyanidin C1

Synonym(s):
Cinnamtannin A1, Proanthocyanidin C1, Procyanidol C1
Empirical Formula (Hill Notation):
C45H38O18
CAS Number:
Molecular Weight:
866.77
Products
Formula Weight
Manufacturer
Product Line
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing