
Norfloxacin Related Compound H

Synonym(s):
7-[4-(Ethoxycarbonyl)piperazin-1-yl]-1-ethyl-6-fluoro-4-oxo-1,4-dihydroquinoline-3-carboxylic acid
Empirical Formula (Hill Notation):
C19H22FN3O5
CAS Number:
Molecular Weight:
391.39
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing