
N-Nitrosodiethanolamine

Synonym(s):
2,2′-(Nitrosoimino)bisethanol, N,N-Diethanolnitrosamine, Bis(2-hydroxyethyl)nitrosamine, NDELA
Empirical Formula (Hill Notation):
C4H10N2O3
CAS Number:
Molecular Weight:
134.13
Products
Brand
Formula Weight
Physical Form
Format
Quality Segment
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing