
Ginsenoside Rg3

Synonym(s):
(3β,12β)-12,20-Dihydroxydammar-24-en-3-yl 2-O-β-D-glucopyranosyl-β-D-glucopyranoside, 20(S)-Ginsenoside-Rg3, Rg3
Empirical Formula (Hill Notation):
C42H72O13
CAS Number:
Molecular Weight:
785.01
Products
Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Research Area
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing