Skip to Content
Merck
Fluoranthene analytical standard

Fluoranthene

Fluoranthene analytical standard
Synonym(s):
Benzo[j,k]fluorene
Empirical Formula (Hill Notation):
C16H10
CAS Number:
Molecular Weight:
202.25
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Format
Application
Markush Class
Markush Group
Package Size
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing