
DOTAP Liposomal Transfection Reagent

Synonym(s):
DOTAP methosulfate, N-(2,3-Dioleoyloxy-1-propyl)trimethylammonium methyl sulfate, Transfection reagent
Empirical Formula (Hill Notation):
C43H83NO8S
CAS Number:
Molecular Weight:
774.19
Products
Formula Weight
Manufacturer
Physical Form
Purity
Quality Segment
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing