Skip to Content
Merck
Cyclohexyl acetate ≥98%, FG

Cyclohexyl acetate

Cyclohexyl acetate ≥98%, FG
Linear Formula:
CH3COOC6H11
CAS Number:
Molecular Weight:
142.20
EC Number:

Products

Product Category
Biological Source
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing