
Aspartame Acesulfame

Synonym(s):
Acesulfame aspartame salt
Empirical Formula (Hill Notation):
C18H23N3O9S
CAS Number:
Molecular Weight:
457.45
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2