
1,3-Bis(isopropylamino)propan-2-ol dihydrochloride

Synonym(s):
3-Bis[(1-methylethyl)amino]propan-2-ol dihydrochloride, Metoprolol impurity M (PhEur) as HCl salt
Empirical Formula (Hill Notation):
C9H22N2O·2HCl
CAS Number:
Molecular Weight:
247.21
Products
Brand
Formula Weight
Manufacturer
Product Line
Quality Segment
Application
Special Grade
Showing 1-1 of 1