
11-Ketoprogesterone

Synonym(s):
4-Pregnene-3,11,20-trione
Empirical Formula (Hill Notation):
C21H28O3
CAS Number:
Molecular Weight:
328.45
Products
Brand
Formula Weight
Physical Form
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing