Merck
HPLC Analysis of Devrinol (Napropamide) Enantiomers on Astec® CHIROBIOTIC® V
HPLC Analysis of Devrinol (Napropamide) Enantiomers on Astec® CHIROBIOTIC® V application for HPLC
Materiały
kolumna analityczna
Numer produktu
Opis
Cennik
Astec® CHIROBIOTIC® V Chiral HPLC Column
5 μm particle size, L × I.D. 25 cm × 4.6 mm
CONDITIONS
column
CHIROBIOTIC V, 25 cm x 4.6 mm I.D., 5 μm particles (11024AST)
mobile phase
[A] 0.1% triethylamine, pH 5.0; [B] THF; (80:20, A:B)
flow rate
1 mL/min
column temp.
25 °C
detector
UV, 254 nm