- Assignment of the gene encoding the catalytic subunit C beta of cAMP-dependent protein kinase to the p36 band on chromosome 1.
Assignment of the gene encoding the catalytic subunit C beta of cAMP-dependent protein kinase to the p36 band on chromosome 1.
Human genetics (1992-03-01)
J Simard, D Bérubé, M Sandberg, K H Grzeschik, R Gagné, V Hansson, T Jahnsen
PMID1551670
ABSTRAKT
A cDNA for the human catalytic subunit (C beta) of cAMP-dependent protein kinase (PKA) has been cloned from a testis cDNA library. In the present study, we have determined the chromosomal localization of this gene using a cDNA for C beta as a probe. Southern blot analysis of genomic DNA from human/mouse cell hybrids revealed that the presence or absence of a 20-kb XbaI fragment, which hybridized with the C beta probe, was concordant with the presence of human chromosome 1. In situ hybridization to metaphase chromosome confirmed the somatic cell hybrid data and regionally mapped the C beta gene of PKA to the p36 band on chromosome 1.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
PKAcβ, active, GST tagged human, PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution