Merck
Inhibitory effects of plant-derived flavonoids and phenolic acids on malonaldehyde formation from ethyl arachidonate.
Journal of agricultural and food chemistry (2003-11-13)
Kwang-Geun Lee, Takayuki Shibamoto, Gary R Takeoka, Sung-Eun Lee, Jeong-Han Kim, Byeoung-Soo Park
ABSTRAKT
The antioxidant activities of naturally occurring plant compounds were measured in a lipid peroxidation system consisting of ethyl arachidonate and Fenton's reagent. Inhibitory effects of 24 plant-derived flavonoids and 5 phenolic acids on malonaldehyde (MA) formation from ethyl arachidonate were examined using gas chromatography (GC) with a nitrogen-phosphorus detector (NPD). Luteolin, which showed the strongest antioxidant activity, inhibited MA formation by 94% and 97% at the levels of 0.5 and 1.0 mM, respectively. The antioxidant activities of the flavones and flavonols decreased in the following order: luteolin > rhamnetin > fisetin > kaempferol > morin > quercetin. Among the flavanones tested, hesperitin, taxifolin, and naringenin exhibited appreciable antioxidant activities (61-84%) at the 1.0 mM level. The inhibitory effect of epigallocatechin gallate (82.5% at the 1.0 mM level) was the strongest among the flavan-3-ols tested. Ferulic acid had the most potent antioxidant activity (74.6% at the 1.0 mM level) of the phenolic acids tested.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Ethyl arachidonate, ≥98.5% (GC), liquid