Merck
MicroRNA-133a alleviates airway remodeling in asthtama through PI3K/AKT/mTOR signaling pathway by targeting IGF1R.
Journal of cellular physiology (2018-08-28)
Youyou Shao, Lei Chong, Peng Lin, Haiyan Li, Lili Zhu, Qiuping Wu, Changchong Li
ABSTRAKT
Asthma is characterized by chronic inflammation, and long-term chronic inflammation leads to airway remodeling. But the potential regulatory mechanism of airway remodeling is not clearly understood, and there is still no effective way to prevent airway remodeling. Present studies have confirmed the role of microRNAs (miRNAs) in the development of disease, which is known as suppressing translation or degradation of messenger RNA (mRNA) at the posttranscriptional stage. In this study, we described the role of miRNA-133a in asthma and demonstrated it in regulating airway remodeling of asthma through the phosphoinositide 3 kinase (PI3K)/protein kinase B (Akt)/mammalian target of rapamycin (mTOR) signaling pathway by targeting IGF-1 receptor (IGF1R). IGF1R helps in mediating the intracellular signaling cascades. Asthmatic mice models were established by sensitization and Ovalbumin challenge. Adenovirus transfer vector carrying miR-133a or miR-133a sponge sequence was used to build the overexpression or downexpression of miR-133a modeling. Real-time polymerase chain reaction and Western blot were used to determine the alterations in the expression of miR-133a and mRNAs and their corresponding proteins. Results showed that miR-133a was downregulated in asthma. Upregulation of miR-133a expression in airway smooth muscle cells in vivo and in vitro could inhibit the activation of PI3K/AKT/mTOR pathway, and reduce the expression of α-smooth muscle actin (α-SMA), indicating that airway remodeling was inhibited. Functional studies based on luciferase reporter revealed miR-133a as a direct target of IGF1R mRNA. In conclusion, these data suggested that miR-133a regulated the expression of α-SMA through PI3K/AKT/mTOR signaling by targeting IGF1R. miR-133a plays a key role in airway remodeling of asthma and may serve as a potential therapeutic target for managing asthmatic airway remodeling.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Anti-phospho-mTOR (Thr2446) Antibody, Upstate®, from rabbit