Skip to Content
MilliporeSigma
V900266
Calcium chloride
Vetec, reagent grade, 96%
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Linear Formula:
CaCl2
CAS Number:
Molecular Weight:
110.98
UNSPSC Code:
12352300
PubChem Substance ID:
EC Number:
233-140-8
MDL number:
Assay:
96%
Grade:
reagent grade
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
grade
reagent grade
vapor pressure
0.01 hPa ( 20 °C), 0.01 mmHg ( 20 °C)
product line
Vetec
assay
96%
form
powder
mp
772 °C (lit.)
SMILES string
[Cl-].[Cl-].[Ca++]
InChI
1S/Ca.2ClH/h;2*1H/q+2;;/p-2
InChI key
UXVMQQNJUSDDNG-UHFFFAOYSA-L
Legal Information
Vetec is a trademark of Merck KGaA, Darmstadt, Germany
Still not finding the right product?
Explore all of our products under Calcium chloride
pictograms
Exclamation mark
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2
Storage Class
11 - Combustible Solids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Visit the Document Library