Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|---|---|
1 ea | Available to ship TODAYfromMILWAUKEE | $144.00 $144.00 |
About This Item
NACRES:
MA.01
UNSPSC Code:
27112105
$144.00
Web-Only Promotion
Available to ship TODAYDetails
Subscriptions are currently unavailable.
Ships Every 4 weeks
material
non-magnetic alloy (tips may be restraightened when bent)
packaging
pack of 1 ea
manufacturer/tradename
Ted Pella 523-C
size
110 mm
General description
Specifically designed for microscopy applications. All are ultra-high precision tools of Swiss manufacture.
1 of 1
This Item | |||
|---|---|---|---|
| material non-magnetic alloy (tips may be restraightened when bent) | material non-magnetic alloy (tips may be restraightened when bent) | material non-magnetic alloy (tips may be restraightened when bent) | material anti-magnetic stainless steel |
| packaging pack of 1 ea | packaging pack of 1 ea | packaging pack of 1 ea | packaging pack of 1 ea |
| size 110 mm | size 110 mm | size 115 mm | size 115 mm |
| manufacturer/tradename Ted Pella 523-C | manufacturer/tradename Ted Pella 525 | manufacturer/tradename Ted Pella 527 | manufacturer/tradename Ted Pella 507-NM |
Still not finding the right product?
Explore all of our products under Tweezers
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.