Skip to Content
MilliporeSigma
Skip To
M4909
Specimen mounts
For Cambridge, Phillips, Etec Instruments
Synonym(s):
Ted Pella 16111, sample container, sample holder, sample mount, sample storage
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
Size/SKUAvailabilityPrice
100 ea
Available to ship TODAYfromMILWAUKEE
$78.00
About This Item
NACRES:
MA.01
UNSPSC Code:
41100000
Material:
aluminum (with lathe finish)
$78.00
Available to ship TODAYDetails
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
material
aluminum (with lathe finish)
pin diam.
3.2 mm
table diam.
12.7 mm
General description
Surface has barely visible rings which help to locate the center.
Compare Similar Items
View Full Comparison
Show Differences
1 of 1
This Item
Z740413Z740414Z740412
material
aluminum (with lathe finish)
material
polyethylene cap, polypropylene container
material
polyethylene cap, polypropylene container
material
polyethylene cap, polypropylene container
pin diam.
3.2 mm
pin diam.
-
pin diam.
-
pin diam.
-
table diam.
12.7 mm
table diam.
-
table diam.
-
table diam.
-
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Visit the Document Library