Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|---|---|
1 ea | Available to ship TODAYfromMILWAUKEE | $21.00 |
About This Item
NACRES:
NB.51
UNSPSC Code:
41121800
$21.00
Available to ship TODAYDetails
manufacturer/tradename
Heathrow Scientific HS6594
General description
Use for counting blood cells, bacterial colonies, drops of liquid or any repetitive event. Tallies up to 9,999. Features a quick-reset knob Lens for easy reading, and finger ring for a secure grip.
1 of 1
This Item | |||
|---|---|---|---|
| manufacturer/tradename Heathrow Scientific HS6594 | manufacturer/tradename - | manufacturer/tradename Heathrow Scientific HS5360A | manufacturer/tradename Heathrow Scientific HS120000 |
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.