Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|---|---|
1 ea | Check Cart for Availability | $10.00 |
About This Item
NACRES:
NB.12
UNSPSC Code:
41123304
Manufacturer/tradename:
Eppendorf® D100 (Global)
$10.00
Check Cart for Availability
manufacturer/tradename
Eppendorf® D100 (Global)
technique(s)
cryopreservation: suitable
size
10 in. × 10 in.
capacity
100 tubes
Legal Information
Eppendorf is a registered trademark of Eppendorf SE
1 of 1
This Item | |||
|---|---|---|---|
| manufacturer/tradename Eppendorf® D100 (Global) | manufacturer/tradename Eppendorf® D64 (Global) | manufacturer/tradename Eppendorf® 6001011410 (Global) | manufacturer/tradename Eppendorf® 6001000911 (Global) |
| size 10 in. × 10 in. | size 8 in. × 8 in. | size - | size - |
| capacity 100 tubes | capacity 64 tubes | capacity - | capacity (max box number per rack: 11 (1 x 11)) |
| technique(s) cryopreservation: suitable | technique(s) cryopreservation: suitable | technique(s) cryopreservation: suitable | technique(s) cryopreservation: suitable |
Still not finding the right product?
Explore all of our products under Eppendorf® Freezer Box Dividers
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.