Skip to Content
MilliporeSigma
2501S
BIOEAZE EZ Top and Aseptic Connecting Devices
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
UNSPSC Code:
12352200
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
description
Designed to Aseptically Transfer of Liquids into or from Bottles by Quick Connects
sterility
sterile; γ-irradiated
General description
Fits 500 mL or 1 L Bottle with 082 Clear C-Flex® Stopper with 38 mm Cap with 1 x 2 ft. length of 3/16" x 3/8" Clear 082 C-Flex® Tubing with Pinch Clamp and 1/4" Female Polycarbonate Quick Connect with Cap
Packaging
Double Polybagged
Legal Information
BIOEAZE is a trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Visit the Document Library