Merck
20757
Hamilton® needles for LT and TLL syringes
Metal Hub (N), needle size 28 ga (bevel tip), needle L 51 mm (2 in.), pkg of 6 ea
Informacje o tej pozycji
UNSPSC Code:
41122001
Material:
stainless steel needle (point style 2)
Needle l:
51 mm (2 in.)
Pomoc techniczna
Pozwól nam pomóc
material
stainless steel needle (point style 2)
needle size
28 ga (bevel tip)
feature
Metal Hub (N)
packaging
pkg of 6 ea
manufacturer/tradename
Hamilton 90028
needle L
51 mm (2 in.)
compatibility
for use with Hamilton LT and TLL
Legal Information
Hamilton is a registered trademark of Hamilton Co.
Still not finding the right product?
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)