Skip to Content
MilliporeSigma

MABE651

Anti-dimethyl Histone H3 (Lys14), clone RM165 Antibody

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Clone:
RM165, monoclonal
Species reactivity:
human, rabbit
Application:
WB, multiplexing
Citations:
-
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


antibody form

purified antibody

Quality Segment

clone

RM165, monoclonal

purified by

using Protein A

species reactivity

human, rabbit

concentration

(Please refer to lot specific datasheet.)

technique(s)

multiplexing: suitable, western blot: suitable

isotype

IgG

UniProt accession no.

General description

A linear peptide corresponding to human Histone H3 dimethylated at Lys14.
~17 kDa observed. Uncharacterized bands may be observed in some lysate(s).

Application

Multiplex Analysis (Mplex): 0.1-0.5 μg/mL from a representative lot was used in Multiplex.

Biochem/physiol Actions

This antibody reacts to Histone H3 dimethylated at Lysine 14 (K14me2). No cross reactivity with monomethylated Lysine 14 (K14me1), trimethylated Lysine 14 (K14me3), or other methylations in Histone H3.

Physical form

Rabbit monoclonal in PBS with 1% BSA and 0.09% sodium azide

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.Note: Variability in freezer temperatures below -20°C may cause glycerol containing solutions to become frozen during storage.

Analysis Note

Evaluated by Western Blotting in recombinant HeLa protein and HeLa acid extract.

Western Blotting Analysis (WB): 0.25 μg/mL of this antibody detected dimethyl Histone H3 (Lys14) in HeLa acid extract.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Storage Class

12 - Non Combustible Liquids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library