Skip to Content
MilliporeSigma
Skip To
1.04015
Glass beads 3 mm
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
Size/SKUAvailabilityPrice
500 g
Estimated to ship TODAY
$109.00
About This Item
NACRES:
NB.51
UNSPSC Code:
41122409
$109.00
Estimated to ship TODAYDetails
Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
storage temp.
no temp limit
Quality Segment
General description
Glass beads can be used to purify, enrich or separate samples of solids, liquids or gases. They offer excellent batch consistency to ensure reliable, reproducible results.
Compare Similar Items
View Full Comparison
Show Differences
1 of 1
This Item
1.040161.040141.04017
storage temp.
no temp limit
storage temp.
no temp limit
storage temp.
no temp limit
storage temp.
no temp limit
Quality Level
100
Quality Level
100
Quality Level
100
Quality Level
100
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Visit the Document Library
Questions
1–2 of 2 Questions  
  1. Are these beads suitable for grinding plant material in a genogrinder for iron quantitation analyses? Also, will they be stable in a strong acidic environment?
    1 answer
    1. This product is composed of soda lime glass, which is more fragile and subject to chipping and cracking. Typically, for tissue grinding purposes, beads made of borosilicate, zirconium, or ceramic are most common and robust. Currently, there are a number of product options available. Please see the link below to review items that may be of interest:
      https://www.sigmaaldrich.com/search/silica-beads?facet=facet_product_category%3Ahomogenizers&focus=products&page=1&perpage=30&sort=relevance&term=silica%20beads&type=product
      Helpful?
  2. What type of glass is this? Borosilicate or soda-lime glass?
    1 answer
    1. These beads are made of soda lime glass.