Merck
TEM-FF100-AU-UL
Sigma-Aldrich
Ultral-Thin Formvar Supported Gold Square Mesh
size 100 mesh, box of 50
About This Item
Kod UNSPSC:
41111761
NACRES:
MA.24
opakowanie
box of 50
rozmiar
100 mesh
Opis ogólny
A 5-6 nm ultral-thin formvar film supported standard square mesh grids. Gold grid, 100 mesh. All of our film is laid on the shiny side of the grid.