Skip to Content
MilliporeSigma
Skip To
SYNC571120
Synthware water jacketed distillation column with removable hose connections
joint: ST/NS 14/20, jacket H 110 mm
Synonym(s):
Synthware C571120, glass distilling column
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
Size/SKUAvailabilityPrice
1 ea
Available to ship TODAYfromMILWAUKEE
$72.20
About This Item
NACRES:
NB.51
UNSPSC Code:
41121800
$72.20
Available to ship TODAYDetails
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
joint
joint: ST/NS14/20
packaging
pkg of 1 ea
manufacturer/tradename
Synthware C571120
jacket H
110 mm
General description
Indentations at the bottom for supporting packing material. Jacket permits the column to be used as a condenser.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Compare Similar Items
View Full Comparison
Show Differences
1 of 1
This Item
SYNC571200SYNC574200SYNC171200
packaging
pkg of 1 ea
packaging
pkg of 1 ea
packaging
pkg of 1 ea
packaging
pkg of 1 ea
jacket H
110 mm
jacket H
200 mm
jacket H
200 mm
jacket H
200 mm
joint
joint: ST/NS14/20
joint
joint: ST/NS14/20
joint
joint: ST/NS24/40
joint
joint: ST/NS14/20
manufacturer/tradename
Synthware C571120
manufacturer/tradename
Synthware C571200
manufacturer/tradename
Synthware C574200
manufacturer/tradename
Synthware C171200
Still not finding the right product?
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Visit the Document Library