Merck
EN 14110: GC Analysis of Methanol Impurity in Biodiesel on Equity-1 after SPME using 85 μm Polyacrylate Fiber
EN 14110: GC Analysis of Methanol Impurity in Biodiesel on Equity-1 after SPME using 85 μm Polyacrylate Fiber suitable for GC, application for SPME
Materials
analytical column
Product No.
Description
Pricing
Equity-1 Capillary GC Column
L × I.D. 30 m × 0.32 mm, df 1.00 μm
SPME fiber
Product No.
Description
Pricing
SPME fiber assembly polyacrylate (PA)
df 85 μm(PA, for use with manual holder, needle size 24 ga
CONDITIONS
sample/matrix
2 g of B100 biodiesel containing 0.01% methanol and 0.0785% 2-propanol in a 4 mL vial
SPME fiber
PK3 SPME ASSY 85UM POLYACRYLATE FS 24GA MAN (WHITE) (57304)
extraction
headspace, 60 °C for 5 min
desorption process
250 °C for 0.75 min
column
Equity-1, 30 m x 0.32 mm I.D., 1.0 μm (28057-U)
oven
50 °C
inj. temp.
250 °C
detector
FID, 200 °C
carrier gas
helium, 30 cm/sec
liner
0.75 mm I.D., SPME type, straight design (unpacked)
Description
Legal Information
null