Merck
HPLC Analysis of Cetirizine Hydrochloride on Purospher® STAR RP-18 endcapped
HPLC Analysis of Cetirizine Hydrochloride on Purospher® STAR RP-18 endcapped application for HPLC
CONDITIONS
column
Purospher® STAR RP-18 endcapped 150 x 4.6 mm (1.51455.0001)
mobile phase
[A] 100mM NaH2PO4 aqueous solution, adjusted to pH 3.7 with H3PO4; [B] acetonitrile; (60:40, A:B)
flow rate
1 mL/min
pressure
1465 psi 101 Bar (101 Bar)
column temp.
30 °C
detector
232 nm cell: 1 μL/10 mm
injection
20 μL
sample
Weigh appropriate amount of Cetirizine, add diluent to obtain 100 μg/mL (100 ppm) concentration.
Description
Legal Information
Purospher is a registered trademark of Merck KGaA, Darmstadt, Germany