Merck
GC Analysis of cis-Chlordane Enantiomers on Astec® CHIRALDEX G-BP
GC Analysis of cis-Chlordane Enantiomers on Astec® CHIRALDEX™ G-BP suitable for GC
Materials
standard
Product No.
Description
Pricing
Pesticide-HC Calibration Standards Kit
analytical standard
α-Chlordane
PESTANAL®, analytical standard
γ-Chlordane
PESTANAL®, analytical standard
EPA 505/525 Update Pesticides Mix B
certified reference material, 500 μg/mL each component in acetone
Chlordane (mixture of isomers)
certified reference material, 1000 μg/mL in isooctane
Chlordane (mixture of isomers)
certified reference material, 500 μg/mL in methanol
Chlordane
analytical standard
Chlordane (mixture of isomers)
certified reference material, 5000 μg/mL in methanol
α-Chlordane solution
certified reference material, 100 μg/mL in hexane
EPA Pesticide Standard Mix B
certified reference material, in hexane: toluene (99:1) (varied)
α-Chlordane solution
10 μg/mL in cyclohexane, PESTANAL®, analytical standard
CONDITIONS
column
Astec CHIRALDEX G-BP, 10 m x 0.25 mm I.D., 0.12 μm (75031AST)
oven
170 °C
inj. temp.
250 °C
detector
FID, 250 °C
carrier gas
helium, 15 psi
injection
0.2 μL, 100:1 split
sample
5 mg/mL in ethanol
peaks 1 & 2: cis-chlordane enantiomers
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIRALDEX is a trademark of Sigma-Aldrich Co. LLC