Merck
GC Analysis of Tall Oil Fatty Acid Methyl Esters on SP®-2380
GC Analysis of Tall Oil Fatty Acid Methyl Esters on SP®-2380 suitable for GC
Materials
analytical column
Product No.
Description
Pricing
SP®-2380 Capillary GC Column
L × I.D. 30 m × 0.25 mm, df 0.20 μm
CONDITIONS
column
SP-2380, 30 m × 0.25 mm I.D., 0.20 μm (24110-U)
oven
170 °C, 4 °C/min to 260 °C (4 min)
inj. temp.
250 °C
detector
FID, 280 °C
carrier gas
helium, 20 cm/sec @ 185°C
injection
2 μL, split 100:1
sample
Tall oil methyl esters in methylene chloride
Description
Legal Information
SP is a registered trademark of Sigma-Aldrich Co. LLC