GC Analysis of PUFA No. 3 Mix (Menhaden Oil Polyunsaturated Fatty Acid (PUFA) Methyl Esters) on SLB®-IL60
CONDITIONS
column
SLB-IL60, 30 m x 0.25 mm I.D., 0.20 μm (29505-U)
oven
170 °C, 1 °C/min to 225 °C
inj. temp.
250 °C
detector
FID, 260 °C
carrier gas
helium, 1.2 mL/min
injection
1.0 μL 100:1 split
liner
4 mm I.D., split/splitless type, single taper wool packed FocusLiner™ design
sample
PUFA No. 3 Mix (47085-U), diluted in 1 mL of hexane
Description
Legal Information
FocusLiner is a trademark of SGE Analytical Science Pty Ltd.
SLB is a registered trademark of Merck KGaA, Darmstadt, Germany