Merck
GC Analysis of FAMEs in Rapeseed Oil (with Conjugated Linoleic Acid (CLA) FAME Isomers Added) on SLB®-IL111
GC Analysis of FAMEs in Rapeseed Oil (with Conjugated Linoleic Acid (CLA) FAME Isomers Added) on SLB®-IL111 suitable for GC
Materials
analytical column
Product No.
Description
Pricing
SP®-2560 Capillary GC Column
L × I.D. 200 m × 0.25 mm, df 0.20 μm
CONDITIONS
column
SLB-IL111, 200 m x 0.25 mm I.D., 0.20 μm (29689-U)
oven
150 °C
inj. temp.
250 °C
detector
FID, 250 °C
carrier gas
hydrogen
injection
1μL, split 50:1
liner
4 mm I.D., split type, cup design
sample
Mix of rapeseed oil FAME isomers and CLA FAME isomers
Description
Analysis Note
Four key conjugated linoleic acid (CLA) FAME isomers are resolved in a mixture with rapeseed oil FAME isomers. The SLB-IL111 provides a different elution pattern compared to SP-2560 (a cyanopropyl siloxane column).
Legal Information
SLB is a registered trademark of Merck KGaA, Darmstadt, Germany