Merck
GC Analysis of Alcoholic Spirit on Equity-1 Coupled to SUPELCOWAX 10 (Column Set 2)
GC Analysis of Alcoholic Spirit on Equity-1 Coupled to SUPELCOWAX 10 (Column Set 2) suitable for GC
Materials
analytical column
Product No.
Description
Pricing
Equity-1 Capillary GC Column
L × I.D. 30 m × 0.32 mm, df 5.00 μm
CONDITIONS
column
Equity-1, 30 m × 0.32 mm I.D., 5.0 μm coupled to SUPELCOWAX 10, 10 m × 0.32 mm I.D., 0.25μm (28062-U)
oven
45 °C (1 min), 10 °C/min to 150 °C, 15 °C/min to 240 °C (2 min)
inj. temp.
250 °C
detector
FID, 250 °C
carrier gas
helium, 36 cm/sec @ 45 °C
injection
1 μL, 11:1 split
sample
alcoholic spirit
Description
Legal Information
null