GC Analysis of a 37-Component FAME Mix on Equity-1 (15 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
Equity-1, 15 m × 0.10 mm I.D., 0.10 μm (28039-U)
oven
100 °C, 50 °C/min to 300 °C (1 min)
inj. temp.
250 °C
detector
FID, 300 °C
carrier gas
hydrogen, 50 cm/sec constant
injection
0.2 μL, 200:1 split
liner
4 mm I.D., split, cup design
sample
Supelco 37-Component FAME Mix (47885-U), analytes at concentrations indicated in methylene chloride
Description
Legal Information
null