Merck
  • [Determination of gomisin A (TJN-101) and its metabolite in rat serum by gas chromatography-mass spectrometry].
[Determination of gomisin A (TJN-101) and its metabolite in rat serum by gas chromatography-mass spectrometry].
Yakugaku zasshi : Journal of the Pharmaceutical Society of Japan (1991-10-01)
Y Matsuzaki, E Ishibashi, S Koguchi, Y Wakui, S Takeda, M Aburada, T Oyama
ABSTRACT
Gomisin A (TJN-101) is one of the lignan components isolated from Schisandra Fruits. A high sensitive and precise method for the determination of TJN-101 and its major metabolite (Met. B) in the rat serum was developed by selected ion monitoring (SIM) with gas chromatography-mass spectrometry (GC/MS) using a fused silica capillary column (SPB-1, Supelco). A 100 microliter serum sample was used for the solid phase extraction. The calibration curves of TJN-101 and Met.B both showed a good linearity between 2.0 and 2000.0 ng/ml. The analytical precision (intra-assay, C.V. less than 4.7%), recoveries (98.4 +/- 10.1%), and detection limit (2 ng/ml) of TJN-101 indicated that this system was suited for the determination of TJN-101 in biological fluid. In case of Met.B, the same results as TJN-101, were obtained. After oral administration of TJN-101 at a dose of 10 mg/kg to male rats, the average values of the maximal serum concentration of TJN-101 and Met.B were 1446.1 +/- 131.8 and 317.4 +/- 18.5 ng/ml, respectively. The serum concentrations of these substances could be monitored sufficiently for 8 h after dosing.
Product Description
Supelco
SPB®-1 Capillary GC Column, L × I.D. 30 m × 0.25 mm, df 0.25 μm