Merck
  • Reconstitution of T Cell Proliferation under Arginine Limitation: Activated Human T Cells Take Up Citrulline
Reconstitution of T Cell Proliferation under Arginine Limitation: Activated Human T Cells Take Up Citrulline
Frontiers in immunology (2017-08-10)
Anke Werner, Miriam Koschke, Nadine Leuchtner, Claudia Luckner-Minden, Alice Habermeier, Johanna Rupp, Christin Heinrich, Roland Conradi, Ellen I Closs, Markus Munder
ABSTRACT
In the tumor microenvironment, arginine is metabolized by arginase-expressing myeloid cells. This arginine depletion profoundly inhibits T cell functions and is crucially involved in tumor-induced immunosuppression. Reconstitution of adaptive immune functions in the context of arginase-mediated tumor immune escape is a promising therapeutic strategy to boost the immunological antitumor response. Arginine can be recycled in certain mammalian tissues from citrulline
Sigma-Aldrich
Anti-ASS1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1
Sigma-Aldrich
Anti-ASL antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution