Merck
  • Room-Temperature Performance of Poly(Ethylene Ether Carbonate)-Based Solid Polymer Electrolytes for All-Solid-State Lithium Batteries.
Room-Temperature Performance of Poly(Ethylene Ether Carbonate)-Based Solid Polymer Electrolytes for All-Solid-State Lithium Batteries.
Scientific reports (2017-12-14)
Yun-Chae Jung, Myung-Soo Park, Duck-Hyun Kim, Makoto Ue, Ali Eftekhari, Dong-Won Kim
ABSTRACT
Amorphous poly(ethylene ether carbonate) (PEEC), which is a copolymer of ethylene oxide and ethylene carbonate, was synthesized by ring-opening polymerization of ethylene carbonate. This route overcame the common issue of low conductivity of poly(ethylene oxide)(PEO)-based solid polymer electrolytes at low temperatures, and thus the solid polymer electrolyte could be successfully employed at the room temperature. Introducing the ethylene carbonate units into PEEC improved the ionic conductivity, electrochemical stability and lithium transference number compared with PEO. A cross-linked solid polymer electrolyte was synthesized by photo cross-linking reaction using PEEC and tetraethyleneglycol diacrylate as a cross-linking agent, in the form of a flexible thin film. The solid-state Li/LiNi
Sigma-Aldrich
1,3-Propylene sulfite, 99%
Sigma-Aldrich
Dibutyltin diacetate, technical grade
Sigma-Aldrich
2-Hydroxy-2-methylpropiophenone, 97%