Merck
Giant coercivity and high magnetic blocking temperatures for N
Nature communications (2017-12-17)
Selvan Demir, Miguel I Gonzalez, Lucy E Darago, William J Evans, Jeffrey R Long
ABSTRACT
Increasing the operating temperatures of single-molecule magnets-molecules that can retain magnetic polarization in the absence of an applied field-has potential implications toward information storage and computing, and may also inform the development of new bulk magnets. Progress toward these goals relies upon the development of synthetic chemistry enabling enhancement of the thermal barrier to reversal of the magnetic moment, while suppressing alternative relaxation processes. Herein, we show that pairing the axial magnetic anisotropy enforced by tetramethylcyclopentadienyl (Cp
Sigma-Aldrich
1,2,3,4-Tetramethyl-1,3-cyclopentadiene, ~85%