Merck
  • Mapping Solvation Environments in Porous Metal-Organic Frameworks with Infrared Chemical Imaging.
Mapping Solvation Environments in Porous Metal-Organic Frameworks with Infrared Chemical Imaging.
The journal of physical chemistry letters (2017-10-13)
Ayanjeet Ghosh, Prabuddha Mukherjee, Sanghamitra Deb, Rohit Bhargava
ABSTRACT
We report here the first mesoscale characterization of solvent environments in the metal-organic framework (MOF) Cu