Merck
Diols, α-Ketols, and Diones as 2
Journal of the American Chemical Society (2016-12-10)
Hiroki Sato, Matthias Bender, Weijie Chen, Michael J Krische
ABSTRACT
The first use of vicinal diols, ketols, or diones as 2
Sigma-Aldrich
TBS-DHG Catalyst, ≥95%