- Synthesis and antineoplastic activity of 3'-azido and 3'-amino analogues of pyrimidine deoxyribonucleoside.
Synthesis and antineoplastic activity of 3'-azido and 3'-amino analogues of pyrimidine deoxyribonucleoside.
Journal of medicinal chemistry (1983-04-01)
T S Lin, W R Mancini
PMID6834387
ABSTRACT
Several new 3'-azido and 3'-amino nucleosides (8, 9, 12, and 13) have been synthesized and their biological activities evaluated. Among them, 3'-amino-2',3'-dideoxycytidine (13) was found to exhibit potent cytotoxic activity against both L1210 and S-180 cells in vitro with an ID50 of 0.7 and 4.0 microM, respectively. Furthermore, 13 has also shown antitumor activity against L1210 tumor bearing mice with a T/C X 100 value of 283.