Merck
  • Preparation of pillar[5]arene-quinoline Langmuir-Blodgett thin films for detection of volatile organic compounds with host-guest principles.
Preparation of pillar[5]arene-quinoline Langmuir-Blodgett thin films for detection of volatile organic compounds with host-guest principles.
The Analyst (2017-08-30)
Ahmed Nuri Kursunlu, Yaser Acikbas, Mustafa Ozmen, Matem Erdogan, Rifat Capan
ABSTRACT
In this study, a novel pillar[5]arene-quinoline (P5-Q) as an organic material is used to fabricate Langmuir-Blodgett (LB) thin films and its organic vapor sensing properties have been investigated. The LB deposition process is characterized by UV-visible spectroscopy, atomic force microscopy (AFM), scanning electron microscopy (SEM) and quartz crystal microbalance (QCM) techniques. The typical frequency shift per layer is obtained as 31.75 Hz per layer and the deposited mass onto a quartz crystal is calculated to be 539.69 ng per layer (2.03 ng mm