Merck
  • Iodinated trihalomethane formation during chloramination of iodate-containing waters in the presence of zero valent iron.
Iodinated trihalomethane formation during chloramination of iodate-containing waters in the presence of zero valent iron.
Water research (2017-08-02)
Ying Xia, Yi-Li Lin, Bin Xu, Chen-Yan Hu, Ze-Chen Gao, Wen-Hai Chu, Nai-Yun Gao
ABSTRACT
Iodide (I
Sigma-Aldrich
2-Iodophenol, 98%