Merck
  • Influence of the penetration enhancer isopropyl myristate on stratum corneum lipid model membranes revealed by neutron diffraction and
Influence of the penetration enhancer isopropyl myristate on stratum corneum lipid model membranes revealed by neutron diffraction and
Biochimica et biophysica acta (2017-01-31)
Adina Eichner, Sören Stahlberg, Stefan Sonnenberger, Stefan Lange, Bodo Dobner, Andreas Ostermann, Tobias E Schrader, Thomas Hauß, Annett Schroeter, Daniel Huster, Reinhard H H Neubert
ABSTRACT
The stratum corneum (SC) provides the main barrier properties in native skin. The barrier function is attributed to the intercellular lipids, forming continuous multilamellar membranes. In this study, SC lipid membranes in model ratios were enriched with deuterated lipids in order to investigate structural and dynamical properties by neutron diffraction and
Sigma-Aldrich
Behenic acid, 99%
Sigma-Aldrich
(2S,3S,4R)-2-Amino-1,3,4-octadecanetriol, AldrichCPR