Merck
  • Phosphorylated HBO1 at UV irradiated sites is essential for nucleotide excision repair.
Phosphorylated HBO1 at UV irradiated sites is essential for nucleotide excision repair.
Nature communications (2017-07-19)
Hiroyuki Niida, Ryoichi Matsunuma, Ryo Horiguchi, Chiharu Uchida, Yuka Nakazawa, Akira Motegi, Koji Nishimoto, Satoshi Sakai, Tatsuya Ohhata, Kyoko Kitagawa, Shinichi Moriwaki, Hideo Nishitani, Ayako Ui, Tomoo Ogi, Masatoshi Kitagawa
ABSTRACT
HBO1, a histone acetyl transferase, is a co-activator of DNA pre-replication complex formation. We recently reported that HBO1 is phosphorylated by ATM and/or ATR and binds to DDB2 after ultraviolet irradiation. Here, we show that phosphorylated HBO1 at cyclobutane pyrimidine dimer (CPD) sites mediates histone acetylation to facilitate recruitment of XPC at the damaged DNA sites. Furthermore, HBO1 facilitates accumulation of SNF2H and ACF1, an ATP-dependent chromatin remodelling complex, to CPD sites. Depletion of HBO1 inhibited repair of CPDs and sensitized cells to ultraviolet irradiation. However, depletion of HBO1 in cells derived from xeroderma pigmentosum patient complementation groups, XPE, XPC and XPA, did not lead to additional sensitivity towards ultraviolet irradiation. Our findings suggest that HBO1 acts in concert with SNF2H-ACF1 to make the chromosome structure more accessible to canonical nucleotide excision repair factors.
Sigma-Aldrich
MISSION® esiRNA, targeting human DDB2
Sigma-Aldrich
MISSION® esiRNA, targeting human XPC
Sigma-Aldrich
BIO, ≥98% (HPLC)
Sigma-Aldrich
Nocodazole, ≥99% (TLC), powder
Sigma-Aldrich
Anti-acetyl-Histone H4 Antibody, serum, Upstate®